Ethyl-8-bromo-6-methylimidazo[1,2-a]pyridine-2-carboxylate 98%
Supplier: Apollo Scientific
OR11401-1G OR11401-5G OR11401-25G
APOSOR11401-1GEA 116 EUR
APOSOR11401-1G APOSOR11401-5G APOSOR11401-25G
Ethyl-8-bromo-6-methylimidazo[1,2-a]pyridine-2-carboxylate 98%
Ethyl 8-bromo-6-methylimidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₁H₁₁BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09475852
CAS Number: 847446-55-3