6-Bromo-4-quinazolone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 6-Bromo-4(3H)-quinazolinone
OR11429-5G OR11429-100G OR11429-25G
APOSOR11429-5GEA 95.7 EUR
APOSOR11429-5G APOSOR11429-100G APOSOR11429-25G
6-Bromo-4-quinazolone 98%
6-Bromo-4-quinazolone
Formula: C₈H₅BrN₂O
MW: 225.04 g/mol
Melting Pt: 272…275 °C
Storage Temperature: Ambient
MDL Number: MFCD01219541
CAS Number: 32084-59-6