6-Bromo-7-methylimidazo[1,2-a]pyridine hydrochloride 98%
Supplier: Apollo Scientific
Warning
OR11461-1G OR11461-5G
APOSOR11461-1GEA 95.7 EUR
APOSOR11461-1G APOSOR11461-5G
6-Bromo-7-methylimidazo[1,2-a]pyridine hydrochloride 98%
6-Bromo-7-methylimidazo[1,2-a]pyridine hydrochloride
Formula: C₈H₈BrClN₂
MW: 247.5200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09475875
CAS Number: 957035-22-2