4-Chloro-o-anisidine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Chloro-2-methoxyaniline
OR11504-5G OR11504-1G
APOSOR11504-5GEA 49.3 EUR
APOSOR11504-5G APOSOR11504-1G
4-Chloro-o-anisidine 98%
4-Chloro-o-anisidine
Formula: C₇H₈ClNO
MW: 157.5975 g/mol
Boiling Pt: 260 °C
Melting Pt: 48…53 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06801386
CAS Number: 93-50-5
EINECS: 202-251-3