2-Chloro-6-nitroanisole 98%
Supplier: Apollo Scientific
Warning
OR11522-25G OR11522-100G
APOSOR11522-25GEA 59.6 EUR
APOSOR11522-25G APOSOR11522-100G
2-Chloro-6-nitroanisole 98%
2-Chloro-6-nitroanisole
Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
Melting Pt: 54…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00065066
CAS Number: 80866-77-9
EINECS: 279-580-4