Ethyl-6-bromopicolinate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 6-bromopyridine-2-carboxylate
OR11544-25G OR11544-5G
APOSOR11544-25GEA 109 EUR
APOSOR11544-25G APOSOR11544-5G
Ethyl-6-bromopicolinate 98%
Ethyl 6-bromopicolinate
Formula: C₈H₈BrNO₂
MW: 230.0600 g/mol
Melting Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD04116913
CAS Number: 21190-88-5