7-Hydroxyflavone 95%
Supplier: Apollo Scientific
Warning
Synonyms: 7-Hydroxy-2-phenylchromen-4-one, 7-Hydroxy-2-phenyl-4-benzopyrone, 7-Hydroxy-2-phenyl-4H-1-benzopyran-4-one, 7-Hydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 7-Hydroxy-2-phenyl-4H-chromen-4-one
OR1155-1G OR1155-25G OR1155-5G
APOSOR1155-1GEA 31.9 EUR
APOSOR1155-1G APOSOR1155-25G APOSOR1155-5G
7-Hydroxyflavone 95%
7-Hydroxyflavone
Formula: C₁₅H₁₀O₃
MW: 238.24 g/mol
Melting Pt: 243…247 °C
Storage Temperature: Ambient
MDL Number: MFCD00006835
CAS Number: 6665-86-7