5-Bromo-N-methylpicolinamide 98%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromo-N-methylpyridine-2-carboxamide
OR11592-250MG OR11592-1G OR11592-100MG
APOSOR11592-250MGEA 43.5 EUR
APOSOR11592-250MG APOSOR11592-1G APOSOR11592-100MG
5-Bromo-N-methylpicolinamide 98%
5-Bromo-N-methylpicolinamide
Formula: C₇H₇BrN₂O
MW: 215.05 g/mol
Melting Pt: 76…77 °C
Storage Temperature: Ambient
MDL Number: MFCD07357421
CAS Number: 845305-87-5