5-Bromovanillin
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-4-hydroxy-5-methoxybenzaldehyde
OR1162-100G OR1162-25G OR1162-500G
APOSOR1162-100GEA 72.5 EUR
APOSOR1162-100G APOSOR1162-25G APOSOR1162-500G
5-Bromovanillin
5-Bromovanillin
Formula: C₈H₇BrO₃
MW: 231.05 g/mol
Boiling Pt: 314
Melting Pt: 164…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00006940
CAS Number: 2973-76-4
EINECS: 221-016-6