2-Amino-3-nitropyridine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Nitropyridin-2-ylamine, 3-Nitropyridin-2-amine, 3-Nitro-2-pyridylamine
OR1163-25G OR1163-5G
APOSOR1163-25GEA 68.3 EUR
APOSOR1163-25G APOSOR1163-5G
2-Amino-3-nitropyridine 98%
2-Amino-3-nitropyridine
Formula: C₅H₅N₃O₂
Melting Pt: 163…167 °C
Flash Pt: 167 °C
Storage Temperature: Ambient
MDL Number: MFCD00006314
CAS Number: 4214-75-9
EINECS: 224-144-0