Ethyl-2-bromo-4-oxazolecarboxylate 98%
Supplier: Apollo Scientific
Warning
OR11662-5G OR11662-1G OR11662-10G
APOSOR11662-5GEA 154 EUR
APOSOR11662-5G APOSOR11662-1G APOSOR11662-10G
Ethyl-2-bromo-4-oxazolecarboxylate 98%
Ethyl 2-bromo-4-oxazolecarboxylate
Formula: C₆H₆BrNO₃
Melting Pt: 49…51 °C
Storage Temperature: Freezer
MDL Number: MFCD09801023
CAS Number: 460081-20-3