1-tert-Butyl-4-piperidone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(tert-Butyl)piperidin-4-one
OR11675-1G OR11675-250MG
APOSOR11675-1GEA 43.5 EUR
APOSOR11675-1G APOSOR11675-250MG
1-tert-Butyl-4-piperidone 98%
1-tert-Butyl-4-piperidone
Formula: C₉H₁₇NO
MW: 155.24 g/mol
Boiling Pt: 93…94 °C
Storage Temperature: Ambient
MDL Number: MFCD03411608
CAS Number: 1465-76-5