4-Methoxy-2-nitrobenzoic acid
Supplier: Apollo Scientific
Warning
Synonyms: 2-Nitro-p-anisic acid, 2-Nitro-4-methoxybenzoic acid
OR11932-1G OR11932-25G OR11932-5G
APOSOR11932-1GEA 21.9 EUR
APOSOR11932-1G APOSOR11932-25G APOSOR11932-5G
4-Methoxy-2-nitrobenzoic acid
4-Methoxy-2-nitrobenzoic acid
Formula: C₈H₇NO₅
MW: 197.15 g/mol
Melting Pt: 196…201 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04972109
CAS Number: 33844-21-2