4-(6-Chloro-4-pyrimidinyl)morpholine
Supplier: Apollo Scientific
Warning
Synonyms: 4-(6-Chloropyrimidin-4-yl)morpholine
OR11965-1G OR11965-5G
APOSOR11965-1GEA 128 EUR
APOSOR11965-1G APOSOR11965-5G
4-(6-Chloro-4-pyrimidinyl)morpholine
4-(6-Chloro-4-pyrimidinyl)morpholine
Formula: C₈H₁₀ClN₃O
MW: 199.64 g/mol
Melting Pt: 149…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00466219
CAS Number: 22177-92-0