2-Amino-5-bromopyridine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromo-2-pyridylamine, 5-Bromopyridin-2-amine
OR12001-100G OR12001-1KG OR12001-250G
APOSOR12001-100GEA 21.9 EUR
APOSOR12001-100G APOSOR12001-1KG APOSOR12001-250G
2-Amino-5-bromopyridine 98%
2-Amino-5-bromopyridine
Formula: C₅H₅BrN₂
Melting Pt: 136…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00006323
CAS Number: 1072-97-5
EINECS: 214-019-9