2-Formyl-6-picoline
Supplier: Apollo Scientific
Warning
Synonyms: 6-Methylpyridine-2-carboxaldehyde, 6-Methyl-2-pyridinecarboxaldehyde
OR12003-25G
APOSOR12003-25GEA 69.6 EUR
APOSOR12003-25G
2-Formyl-6-picoline
2-Formyl-6-picoline
Formula: C₇H₇NO
MW: 121.14 g/mol
Melting Pt: 31…33 °C
Flash Pt: 68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006291
CAS Number: 1122-72-1
EINECS: 214-359-8