3-Cyclopentene-1-carboxylic acid 97%
Supplier: Apollo Scientific
Warning
Synonyms: cyclopent-3-ene-1-carboxylic acid, Cyclopent-3-enecarboxylic acid
OR12005-25G OR12005-100G OR12005-5G
APOSOR12005-25GEA 29.5 EUR
APOSOR12005-25G APOSOR12005-100G APOSOR12005-5G
3-Cyclopentene-1-carboxylic acid 97%
3-Cyclopentene-1-carboxylic acid
Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 215 °C
Density: 1.084 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00101764
CAS Number: 7686-77-3