2-Bromo-4-methylimidazole
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-4-methyl-1H-imidazole
OR1205-1G OR1205-5G OR1205-10G
APOSOR1205-1GEA 56.7 EUR
APOSOR1205-1G APOSOR1205-5G APOSOR1205-10G
2-Bromo-4-methylimidazole
2-Bromo-4-methylimidazole
Formula: C₄H₅BrN₂
MW: 161 g/mol
Melting Pt: 110…114 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659904
CAS Number: 23328-88-3