6,8-Dibromo-2-methyl-3-quinolinol
Supplier: Apollo Scientific
OR12070-10G OR12070-1G OR12070-5G
APOSOR12070-10GEA 1350 EUR
APOSOR12070-10G APOSOR12070-1G APOSOR12070-5G
6,8-Dibromo-2-methyl-3-quinolinol
6,8-Dibromo-2-methyl-3-quinolinol
Formula: C₁₀H₇Br₂NO
MW: 316.99 g/mol
Melting Pt: 252 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03617848
CAS Number: 59869-01-1