Methyl-2-aminothiazole-4-carboxylate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2-aminothiazole-4-carboxylate
OR1210-25G OR1210-100G OR1210-500G
APOSOR1210-25GEA 23.2 EUR
APOSOR1210-25G APOSOR1210-100G APOSOR1210-500G
Methyl-2-aminothiazole-4-carboxylate 98%
Methyl 2-aminothiazole-4-carboxylate
Formula: C₅H₆N₂O₂S
MW: 158.1800 g/mol
Melting Pt: 171…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00622441
CAS Number: 118452-04-3