Methyl-4-(2-bromoethoxy)benzenecarboxylate
Supplier: Apollo Scientific
Warning
OR12127-1G OR12127-5G
APOSOR12127-1GEA 36.4 EUR
APOSOR12127-1G APOSOR12127-5G
Methyl-4-(2-bromoethoxy)benzenecarboxylate
Methyl-4-(2-bromoethoxy)benzenecarboxylate
Formula: C₁₀H₁₁BrO₃
MW: 259.11 g/mol
Melting Pt: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD01877551
CAS Number: 56850-91-0