4-(2-Oxo-1-imidazolidinyl)benzenecarbonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 4-(2-Oxoimidazolidin-1-yl)benzonitrile
OR12158-1G OR12158-500MG
APOSOR12158-1GEA 69.6 EUR
APOSOR12158-1G APOSOR12158-500MG
4-(2-Oxo-1-imidazolidinyl)benzenecarbonitrile
4-(2-Oxo-1-imidazolidinyl)benzenecarbonitrile
Formula: C₁₀H₉N₃O
MW: 187.2 g/mol
Melting Pt: 179…181 °C
Storage Temperature: Ambient
MDL Number: MFCD08689735
CAS Number: 144655-81-2