4-(2-Pyridinyl)-1H-pyrazol-5-amine
Supplier: Apollo Scientific
Warning
Synonyms: 5-Amino-4-(pyridin-2-yl)-1H-pyrazole
OR12160-1G OR12160-500MG
APOSOR12160-1GEA 94.4 EUR
APOSOR12160-1G APOSOR12160-500MG
4-(2-Pyridinyl)-1H-pyrazol-5-amine
4-(2-Pyridinyl)-1H-pyrazol-5-amine
Formula: C₈H₈N₄
MW: 160.18 g/mol
Melting Pt: 200…202 °C
Storage Temperature: Ambient
MDL Number: MFCD03617434
CAS Number: 493038-87-2