4-Amino-2-bromopyrimidine-5-carbonitrile 95+%
Supplier: Apollo Scientific
Warning
OR1217-1G OR1217-5G
APOSOR1217-1GEA 46.4 EUR
APOSOR1217-1G APOSOR1217-5G
4-Amino-2-bromopyrimidine-5-carbonitrile 95+%
4-Amino-2-bromopyrimidine-5-carbonitrile
Formula: C₅H₃BrN₄
MW: 199.01 g/mol
Melting Pt: 254 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00173661
CAS Number: 94741-70-5