ω-Bromo-2,4-dichloroacetophenone 95%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Bromo-1-(2,4-dichlorophenyl)ethan-1-one, 2,4-Dichlorophenacyl bromide, α-Bromo-2,4-dichloroacetophenone, 2-Bromo-2',4'-dichloroacetophenone, 2-Bromo-1-(2,4-dichlorophenyl)ethanone
OR12270-5G OR12270-100G OR12270-25G
APOSOR12270-5GEA 49.3 EUR
APOSOR12270-5G APOSOR12270-100G APOSOR12270-25G
ω-Bromo-2,4-dichloroacetophenone 95%
ω-Bromo-2,4-dichloroacetophenone
Formula: C₈H₅BrCl₂O
MW: 267.94 g/mol
Melting Pt: 25…29 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00053005
CAS Number: 2631-72-3
EINECS: 220-116-7
UN: 1759
ADR: 8,III