2,4-Dibromo-1H-imidazole 98%
Supplier: Apollo Scientific
Warning
OR1229-1G OR1229-250MG
APOSOR1229-1GEA 132 EUR
APOSOR1229-1G APOSOR1229-250MG
2,4-Dibromo-1H-imidazole 98%
2,4-Dibromo-1H-imidazole
Formula: C₃H₂Br₂N₂
Melting Pt: 193…198 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08741407
CAS Number: 64591-03-3