4-Bromooxindole
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromooxindole, 4-Bromo-2,3-dihydro-1H-indol-2-one, 4-Bromoindolin-2-one
OR1230-5G
APOSOR1230-5GEA 84.1 EUR
APOSOR1230-5G
4-Bromooxindole
4-Bromooxindole
Formula: C₈H₆BrNO
MW: 212.05 g/mol
Melting Pt: 202…208 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659911
CAS Number: 99365-48-7