Benzo[b]thiophene-7-carboxaldehyde 97%
Supplier: Apollo Scientific
Warning
Synonyms: Benzo[b]thiophene-7-carbaldehyde
OR12310-1G OR12310-250MG
APOSOR12310-1GEA 0 EUR
APOSOR12310-1G APOSOR12310-250MG
Benzo[b]thiophene-7-carboxaldehyde 97%
Benzo[b]thiophene-7-carboxaldehyde
Formula: C₉H₆OS
MW: 162.2083 g/mol
Melting Pt: 42…44 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08572207
CAS Number: 10134-91-5
UN: 2811
ADR: 6.1 III