N-Methyl-1-(5-methyl-2-thienyl)methanamine hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: 2-Methyl-5-[(methylamino)methyl]thiophene hydrochloride
OR12379-1G OR12379-250MG OR12379-100MG OR12379-500MG
APOSOR12379-1GEA 267 EUR
APOSOR12379-1G APOSOR12379-250MG APOSOR12379-100MG APOSOR12379-500MG
N-Methyl-1-(5-methyl-2-thienyl)methanamine hydrochloride
N-Methyl-1-(5-methyl-2-thienyl)methanamine hydrochloride
Formula: C₇H₁₂ClNS
MW: 177.7 g/mol
Melting Pt: 168…169 °C
Storage Temperature: Ambient
MDL Number: MFCD07106798
CAS Number: 912569-78-9