1-{2-[(5-Chloro-2-pyridinyl)amino]-1,3-thiazol-5-yl}-1-ethanone
Supplier: Apollo Scientific
Synonyms: 5-Acetyl-2-[(5-chloropyridin-2-yl)amino]-1,3-thiazole
OR12486-1G OR12486-5G
APOSOR12486-1GEA 538 EUR
APOSOR12486-1G APOSOR12486-5G
1-{2-[(5-Chloro-2-pyridinyl)amino]-1,3-thiazol-5-yl}-1-ethanone
1-{2-[(5-Chloro-2-pyridinyl)amino]-1,3-thiazol-5-yl}-1-ethanone
Formula: C₁₀H₈ClN₃OS
MW: 253.71 g/mol
Melting Pt: 250…252 °C
Storage Temperature: Ambient
MDL Number: MFCD09817445
CAS Number: 952183-72-1