4-Methoxy-o-toluidine 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Methoxy-2-methylaniline, m-Cresidine, 2-Methyl-p-anisidine, 1-Amino-4-methoxy-2-methylbenzene, 2-Amino-5-methoxytoluene, 4-Amino-3-methylanisole
OR12498-100G
OR12498-25G
APOSOR12498-100GEA
74.1
EUR
APOSOR12498-100G
APOSOR12498-25G
4-Methoxy-o-toluidine 98%
4-Methoxy-o-toluidine
Formula:
C₈H₁₁NO Boiling Pt: 248…249 °C Melting Pt: 13…14 °C Density: 1.065 g/cm³ (25 °C) Flash Pt: 124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007735 CAS Number: 102-50-1 EINECS: 203-036-7 |