2-(3-Pyridyl)benzaldehyde 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(Pyridin-3-yl)benzaldehyde
OR12527-1G
OR12527-250MG
APOSOR12527-1GEA
79.9
EUR
APOSOR12527-1G
APOSOR12527-250MG
2-(3-Pyridyl)benzaldehyde 95%
2-(3-Pyridyl)benzaldehyde
Formula:
C₁₂H₉NO MW: 183.2100 g/mol Melting Pt: 67…68 °C Storage Temperature: Ambient |
MDL Number:
MFCD02684097 CAS Number: 176690-44-1 |