1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane 98%
Supplier: Apollo Scientific
Danger
Synonyms: 1,3-Dichloro-1,1,3,3-tetrakis(isopropyl)disiloxane
OR1258-100G OR1258-25G
APOSOR1258-100GEA 248 EUR
APOSOR1258-100G APOSOR1258-25G
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane 98%
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane
Formula: C₁₂H₂₈Cl₂OSi₂
Boiling Pt: 70 °C/0.5mmHg
Density: 0.986 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009655
CAS Number: 69304-37-6
UN: 2987
ADR: 8,II