4-(4-Chlorophenoxy)benzaldehyde
Supplier: Apollo Scientific
Warning
OR12612-1G OR12612-5G
APOSOR12612-1GEA 33.5 EUR
APOSOR12612-1G APOSOR12612-5G
4-(4-Chlorophenoxy)benzaldehyde
4-(4-Chlorophenoxy)benzaldehyde
Formula: C₁₃H₉ClO₂
MW: 232.67 g/mol
Melting Pt: 54…58 °C
Storage Temperature: Ambient
MDL Number: MFCD01631895
CAS Number: 61343-99-5