(1S,4R)-4-Isopropenyl-1-methylcyclohex-2-en-1-ol 98%
Supplier: Apollo Scientific
Warning
Synonyms: (1S,4R)-1-Methyl-4-(prop-1-en-2-yl)cyclohex-2-en-1-ol, (1S,4R)-1-Methyl-4-(prop-1-en-2-yl)cyclohex-2-enol, cis-Isolimonenol
OR12641-1G OR12641-5G OR12641-25G
APOSOR12641-1GEA 21.9 EUR
APOSOR12641-1G APOSOR12641-5G APOSOR12641-25G
(1S,4R)-4-Isopropenyl-1-methylcyclohex-2-en-1-ol 98%
(1S,4R)-4-Isopropenyl-1-methylcyclohex-2-en-1-ol
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 81…83 °C/7mmHg
Melting Pt: 18 °C
Density: 0.9398 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD08460036
CAS Number: 22972-51-6