4-(2-Pyridyl)benzaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 4-Pyridin-2-yl-benzaldehyde, 4-(Pyridin-2-yl)benzaldehyde
OR12648-1G OR12648-250MG
APOSOR12648-1GEA 24.8 EUR
APOSOR12648-1G APOSOR12648-250MG
4-(2-Pyridyl)benzaldehyde
4-(2-Pyridyl)benzaldehyde
Formula: C₁₂H₉NO
MW: 183.21 g/mol
Melting Pt: 51…53 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD01863537
CAS Number: 127406-56-8