2-(4-Methoxyphenyl)-4,6-bis(trichloromethyl)-1,3,5-triazine
Supplier: Apollo Scientific
Synonyms: 4,6-Bis(trichloromethyl)-2-(4-methoxyphenyl)-1,3,5-triazine
OR12656-25G
APOSOR12656-25GEA 115 EUR
APOSOR12656-25G
2-(4-Methoxyphenyl)-4,6-bis(trichloromethyl)-1,3,5-triazine
2-(4-Methoxyphenyl)-4,6-bis(trichloromethyl)-1,3,5-triazine
Formula: C₁₂H₇Cl₆N₃O
MW: 421.91 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00521486
CAS Number: 3584-23-4