3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carboxylic acid
Supplier: Apollo Scientific
Warning
Synonyms: 3-tert-Butyl-1-methylpyrazole-5-carboxylic Acid
OR1280-10G OR1280-1G OR1280-5G
APOSOR1280-10GEA 0 EUR
APOSOR1280-10G APOSOR1280-1G APOSOR1280-5G
3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carboxylic acid
3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carboxylic acid
Formula: C₉H₁₄N₂O₂
MW: 182.22 g/mol
Melting Pt: 151…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00173812
CAS Number: 175277-11-9