Benzyl bromide 99%
Supplier: Apollo Scientific
Danger
Synonyms: alpha-Bromotoluene
OR12905-100G OR12905-25G
APOSOR12905-100GEA 43.5 EUR
APOSOR12905-100G APOSOR12905-25G
Benzyl bromide 99%
Benzyl bromide
Formula: (C₆H₅)CH₂Br
MW: 171.04 g/mol
Boiling Pt: 198…199 °C
Melting Pt: –3…–1 °C
Density: 1.438 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00000172
CAS Number: 100-39-0
EINECS: 202-847-3
UN: 1737
ADR: 6.1+8 II
Merck Index: 13,01130