2-Chloroethyl phenyl sulphone 98%
Supplier: Apollo Scientific
Danger
Synonyms: [(2-Chloroethyl)sulphonyl]benzene
OR1291-100G OR1291-25G OR1291-5G
APOSOR1291-100GEA 702 EUR
APOSOR1291-100G APOSOR1291-25G APOSOR1291-5G
2-Chloroethyl phenyl sulphone 98%
2-Chloroethyl phenyl sulphone
Formula: C₈H₉ClO₂S
MW: 204.68 g/mol
Boiling Pt: 168…174 °C/7mmHg
Melting Pt: 52…55 °C
Storage Temperature: Ambient
MDL Number: MFCD00025047
CAS Number: 938-09-0
EINECS: 213-337-5