4-Hydroxy-3-methoxybenzylamine hydrochloride 98%
Supplier: Apollo Scientific
Warning
Synonyms: Vanillylamine HCl, 4-(Aminomethyl)-2-methoxyphenol hydrochloride
OR13027-25G OR13027-10G
APOSOR13027-25GEA 24.8 EUR
APOSOR13027-25G APOSOR13027-10G
4-Hydroxy-3-methoxybenzylamine hydrochloride 98%
4-Hydroxy-3-methoxybenzylamine hydrochloride
Formula: C₈H₁₂ClNO₂
MW: 189.64 g/mol
Melting Pt: 219…221 °C
Storage Temperature: Ambient
MDL Number: MFCD00012864
CAS Number: 7149-10-2
EINECS: 230-468-3