Homovanillic acid 99%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(4-Hydroxy-3-methoxyphenyl)acetic acid, (4-Hydroxy-3-methoxyphenyl)acetic acid
OR13029-25G OR13029-5G
APOSOR13029-25GEA 122 EUR
APOSOR13029-25G APOSOR13029-5G
Homovanillic acid 99%
Homovanillic acid
Formula: (C₆H₃(OH)(OCH₃))CH₂COOH
MW: 182.18 g/mol
Boiling Pt: 339
Melting Pt: 140…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00004350
CAS Number: 306-08-1
Merck Index: 13,04761