4-Iodobiphenyl 98%

Supplier: Apollo Scientific
Danger

APOSOR13030-25GEA 62.6 EUR
APOSOR13030-25G
4-Iodobiphenyl 98%
4-Iodobiphenyl
Formula: C₁₂H₉I
MW: 280,11 g/mol
Boiling Pt: 320 °C
Melting Pt: 110…114 °C
Storage Temperature: Refrigerator
EINECS: 216-469-1
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR