1-Bromo-4-propylbenzene
Supplier: Apollo Scientific
Warning
Synonyms: (4-Bromophenyl)propane, 1-Bromo-4-n-propylbenzene
OR13034-100G OR13034-250G OR13034-25G
APOSOR13034-100GEA 39.3 EUR
APOSOR13034-100G APOSOR13034-250G APOSOR13034-25G
1-Bromo-4-propylbenzene
1-Bromo-4-propylbenzene
Formula: C₉H₁₁Br
MW: 199.09 g/mol
Boiling Pt: 225 °C
Density: 1.286 g/cm³ (25 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00012456
CAS Number: 588-93-2