4-Nitro-2,6-xylenol
Supplier: Apollo Scientific
Warning
Synonyms: 2-Hydroxy-5-nitro-3-xylene, 2,6-Dimethyl-4-nitrophenol
OR13035-25G
APOSOR13035-25GEA 82.8 EUR
APOSOR13035-25G
4-Nitro-2,6-xylenol
4-Nitro-2,6-xylenol
Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: 168 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007339
CAS Number: 2423-71-4
EINECS: 219-353-9