2,5-Dibromo-3-picoline
Supplier: Apollo Scientific
Warning
OR13063-100G OR13063-25G
APOSOR13063-100GEA 63 EUR
APOSOR13063-100G APOSOR13063-25G
2,5-Dibromo-3-picoline
2,5-Dibromo-3-picoline
Formula: C₆H₅Br₂N
Melting Pt: 43…47 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093085
CAS Number: 3430-18-0
UN: 2811
ADR: 6.1,III