3-Iodo-4-nitroanisole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Iodo-4-methoxy-1-nitrobenzene
OR13079-25G OR13079-5G OR13079-1G
APOSOR13079-25GEA 190 EUR
APOSOR13079-25G APOSOR13079-5G APOSOR13079-1G
3-Iodo-4-nitroanisole 98%
3-Iodo-4-nitroanisole
Formula: C₇H₆INO₃
MW: 279.03 g/mol
Melting Pt: 68…73 °C
Storage Temperature: Ambient
MDL Number: MFCD01320682
CAS Number: 214279-40-0
UN: 2811
ADR: 6.1,III