4-(4-Methoxybenzoyl)piperidine hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: (4-Methoxyphenyl)(piperidin-4-yl)methanone hydrochloride
OR1309-1G OR1309-5G
APOSOR1309-1GEA 114 EUR
APOSOR1309-1G APOSOR1309-5G
4-(4-Methoxybenzoyl)piperidine hydrochloride
4-(4-Methoxybenzoyl)piperidine hydrochloride
Formula: C₁₃H₁₈ClNO₂
MW: 255.7405 g/mol
Melting Pt: 251…254 °C
Storage Temperature: Ambient
MDL Number: MFCD00114817
CAS Number: 25519-82-8
UN: 2811
ADR: 6.1 III