1,3,5-Triethynylbenzene 98+%
Supplier: Apollo Scientific
Danger
OR13102-1G OR13102-5G OR13102-10G
APOSOR13102-1GEA 21.9 EUR
APOSOR13102-1G APOSOR13102-5G APOSOR13102-10G
1,3,5-Triethynylbenzene 98+%
1,3,5-Triethynylbenzene
Formula: C₁₂H₆
MW: 150.18 g/mol
Melting Pt: 105…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00068621
CAS Number: 7567-63-7
UN: 1325
ADR: 4.1,III