6-Cyanonicotinic acid 97%
Supplier: Apollo Scientific
Warning
OR13130-1G OR13130-250MG
APOSOR13130-1GEA 109 EUR
APOSOR13130-1G APOSOR13130-250MG
6-Cyanonicotinic acid 97%
6-Cyanonicotinic acid
Formula: C₇H₄N₂O₂
MW: 148.12 g/mol
Melting Pt: 187…194 °C
Storage Temperature: Ambient
MDL Number: MFCD01318135
CAS Number: 70165-31-0